Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAT1 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP232274AF647
Description
FAT1 Polyclonal antibody specifically detects FAT1 in Human, Mouse samples. It is validated for Western BlotSpecifications
FAT1 | |
Polyclonal | |
Western Blot | |
FAT tumor suppressor 1, FAT tumor suppressor homolog 1 (Drosophila) | |
The immunogen this antibody was made to, maps to a region between residue 4450 to 4500 of human FAT Tumor Suppressor Homolog 1 using the numbering given in entry NP_005236.2 (GeneID 2195). | |
0.1 mL | |
Signal Transduction | |
2195 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction