Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAT1 Monoclonal Antibody (1634CT464.1.9)
GREENER_CHOICE

Catalog No. PIMA537702
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537702 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537702 Supplier Invitrogen™ Supplier No. MA537702
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FAT1 Monoclonal Antibody for Western Blot

This gene is an ortholog of the Drosophila fat gene, which encodes a tumor suppressor essential for controlling cell proliferation during Drosophila development. The gene product is a member of the cadherin superfamily, a group of integral membrane proteins characterized by the presence of cadherin-type repeats. In addition to containing 34 tandem cadherin-type repeats, the gene product has five epidermal growth factor (EGF)-like repeats and one laminin A-G domain. This gene is expressed at high levels in a number of fetal epithelia. Its product probably functions as an adhesion molecule and/or signaling receptor, and is likely to be important in developmental processes and cell communication. Transcript variants derived from alternative splicing and/or alternative promoter usage exist, but they have not been fully described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAT1
Applications Western Blot
Classification Monoclonal
Clone 1634CT464.1.9
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene FAT1
Gene Accession No. Q14517
Gene Alias 2310038E12Rik; AU023433; Cadherin family member 7; cadherin FAT1 isoform +12; cadherin ME5; cadherin-related family member 8; cadherin-related tumor suppressor homolog; CDHF7; CDHR8; FAT; fat 1 cadherin; FAT atypical cadherin 1; FAT tumor suppressor 1; FAT tumor suppressor homolog 1; FAT tumor suppressor homolog 1 (Drosophila); FAT1; Fath; hFat1; ME5; mFat1; Protein fat homolog; protocadherin Fat 1; Protocadherin Fat 1, nuclear form
Gene Symbols FAT1
Host Species Mouse
Immunogen recombinant protein between 4203-4539 amino acids from human FAT1.
Purification Method Protein G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2195
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.