Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554910
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554910 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554910 Supplier Invitrogen™ Supplier No. PA554910
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NFQRALRNIL GVRRNDIQIV SLQSSEPHPH LDVLLFVEKP GSAQISTKQL LHKINSSVTD IEEIIGVRIL NVFQKLCAGL DCPWKFCDEK VSVDESVMST HSTARLSFVT PRHHRAAVCL CKEGRCP Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 90%.

This gene is an ortholog of the Drosophila fat gene, which encodes a tumor suppressor essential for controlling cell proliferation during Drosophila development. The gene product is a member of the cadherin superfamily, a group of integral membrane proteins characterized by the presence of cadherin-type repeats. In addition to containing 34 tandem cadherin-type repeats, the gene product has five epidermal growth factor (EGF)-like repeats and one laminin A-G domain. This gene is expressed at high levels in a number of fetal epithelia. Its product probably functions as an adhesion molecule and/or signaling receptor, and is likely to be important in developmental processes and cell communication. Transcript variants derived from alternative splicing and/or alternative promoter usage exist, but they have not been fully described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAT1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAT1
Gene Accession No. Q14517
Gene Alias 2310038E12Rik; AU023433; Cadherin family member 7; cadherin FAT1 isoform +12; cadherin ME5; cadherin-related family member 8; cadherin-related tumor suppressor homolog; CDHF7; CDHR8; FAT; fat 1 cadherin; FAT atypical cadherin 1; FAT tumor suppressor 1; FAT tumor suppressor homolog 1; FAT tumor suppressor homolog 1 (Drosophila); FAT1; Fath; hFat1; ME5; mFat1; Protein fat homolog; protocadherin Fat 1; Protocadherin Fat 1, nuclear form
Gene Symbols FAT1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAT1. Recombinant protein control fragment (Product #RP-101000).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2195
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.