Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAT2 Antibody (8C5), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP254371MFV450
Description
FAT2 Monoclonal antibody specifically detects FAT2 in Human samples. It is validated for Flow CytometrySpecifications
FAT2 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
2196 | |
Store at 4C in the dark. | |
IgG1 κ |
Flow Cytometry | |
8C5 | |
Flow Cytometry | |
protocadherin Fat 2, CDHF8, CDHR9, FAT tumor suppressor homolog 2 (Drosophila), HFAT2, MEGF1 | |
Purified recombinant human FAT2 protein. (Uniprot: Q9NYQ8) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction