Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAT2 Antibody (8C5), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP253235200UG
Description
FAT2 Monoclonal antibody specifically detects FAT2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, SDS-Page.Specifications
FAT2 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, SDS-Page | |
protocadherin Fat 2, CDHF8, CDHR9, FAT tumor suppressor homolog 2 (Drosophila), HFAT2, MEGF1 | |
Mouse | |
480 kDa | |
200 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), SDS-Page | |
8C5 | |
Unconjugated | |
10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
FAT2 | |
Purified recombinant human FAT2 fusion protein | |
Protein A or G purified | |
RUO | |
2196 | |
Store at 4C. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction