Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAT2 Antibody (8C5), PE/Cy5.5, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP254371PECY55
Description
FAT2 Monoclonal specifically detects FAT2 in Human samples. It is validated for Flow Cytometry.Specifications
| FAT2 | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS with 0.05% Sodium Azide | |
| FAT2 | |
| Purified recombinant human FAT2 protein. (Uniprot: Q9NYQ8) | |
| 0.1 mL | |
| 2196.0 | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| 8C5 | |
| Flow Cytometry | |
| protocadherin Fat 2, CDHF8, CDHR9, FAT tumor suppressor homolog 2 (Drosophila), HFAT2, MEGF1 | |
| Mouse | |
| Protein A or G purified | |
| Primary | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction