Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAT3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5145082
Description
Immunogen sequence: PSFTQSQYSF TIAEDTAIGS TVDTLRILPS QNVWFSTVNG ERPENNKGGV FVIEQETGTI KLDKRLDRET SPAFHFKVAA TIPLDKVDIV FTVDV Antigen sequence identity: Mouse 88%; Rat 88%.
FAT1, FAT2, FAT3 and FAT4 are human homologs of Drosophila Fat, which is involved in tumor suppression and planar cell polarity (PCP). The atypical cadherin Fat3 ensures that retinal amacrine cells (ACs) develop this unipolar morphology. Fat3 expression was restricted to the nervous system, for example in the brain, it is expressed in a variety of regions and axon fascicles. However, its strongest expression was observed in the olfactory bulb and retina.
Specifications
FAT3 | |
Polyclonal | |
Unconjugated | |
FAT3 | |
9430076A06Rik; Cadherin family member 15; cadherin-related family member 10; CDHF15; CDHR10; D430038H04Rik; FAT atypical cadherin 3; FAT tumor suppressor homolog 3; FAT tumor suppressor homolog 3 (Drosophila); Fat3; Fta3 protein; Gm1132; Gm510; hFat3; KIAA1989; MFAT3F; protocadherin Fat 3; protocadherin Fat3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
120114 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TDW7 | |
FAT3 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 2706-2800. Recombinant protein control fragment (Product #RP-110211). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction