Learn More
Invitrogen™ FAT3 Polyclonal Antibody

Description
The antibody detects endogenous levels of total FAT3 protein.
Specifications
Specifications
| Antigen | FAT3 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.7 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | FAT3 |
| Gene Accession No. | Q8TDW7 |
| Gene Alias | 9430076A06Rik; Cadherin family member 15; cadherin-related family member 10; CDHF15; CDHR10; D430038H04Rik; FAT atypical cadherin 3; FAT tumor suppressor homolog 3; FAT tumor suppressor homolog 3 (Drosophila); Fat3; Fta3 protein; Gm1132; Gm510; hFat3; KIAA1989; MFAT3F; protocadherin Fat 3; protocadherin Fat3 |
| Gene Symbols | FAT3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.