Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAT4 Polyclonal Antibody
FAT4 Polyclonal Antibody
Supplier: Thermo Scientific PA550587
Description
FAT4 Polyclonal Antibody for IHC (P)

Specifications
FAT4 | |
Polyclonal | |
Unconjugated | |
FAT4 | |
6030410K14Rik; 9430004M15; Cadherin family member 14; cadherin-related family member 11; CDHF14; CDHR11; FAT atypical cadherin 4; FAT tumor suppressor homolog 4; FAT tumor suppressor homolog 4 (Drosophila); FAT4; Fatj; FAT-J; fat-like cadherin protein FAT-J; hFat4; HKLLS2; Nbla00548; protocadherin Fat 4; putative protein product of Nbla00548; RGD1564291; VMLDS2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79633 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.7 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q6V0I7 | |
FAT4 | |
Synthetic peptide corresponding to residues near the C terminal of human FAT tumor suppressor homolog 4 (Drosophila). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction