Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAT4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562694
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562694 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562694 Supplier Invitrogen™ Supplier No. PA562694
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FDDPDNIPPY GDDMTVRKQP EGNPKPDIIE RENPYLIYDE TDIPHNSETI PSAPLASPEQ EIEHYDIDNA SSIAPSDADI IQHYKQFRSH T Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

FAT1, FAT2, FAT3 and FAT4 are human homologs of Drosophila Fat, which is involved in tumor suppression and planar cell polarity (PCP). FAT4 undergo the first proteolytic cleavage by Furin and are predicted to undergo the second cleavage by secretase to release intracellular domain (ICD). FAT4 directly interacts with MPDZ/MUPP1 to recruit membrane associated guanylate kinase MPP5/PALS1. FAT4 is involved in the maintenance of PCP and inhibition of cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAT4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAT4
Gene Accession No. Q6V0I7
Gene Alias 6030410K14Rik; 9430004M15; Cadherin family member 14; cadherin-related family member 11; CDHF14; CDHR11; FAT atypical cadherin 4; FAT tumor suppressor homolog 4; FAT tumor suppressor homolog 4 (Drosophila); FAT4; Fatj; FAT-J; fat-like cadherin protein FAT-J; hFat4; HKLLS2; Nbla00548; protocadherin Fat 4; putative protein product of Nbla00548; RGD1564291; VMLDS2
Gene Symbols FAT4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAT4. Recombinant protein control fragment (Product #RP-101703).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79633
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.