Learn More
FATE1 Antibody (OTI1A1), Alexa Fluor™ 488, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP272310AF488
Catalog No. NB272310A48
Only null left
Add to Cart
Description
Description
FATE1 Monoclonal specifically detects FATE1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
FATE1
Monoclonal
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
FATE1
Full length recombinant protein of human FATE1 (NP_149076) produced in HEK293T cells.
0.1 mL
89885.0
Store at 4°C in the dark.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
OTI1A1
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Cancer/testis antigen 43, CT43BJ-HCC-2 antigen, FATEfetal and adult testis expressed transcript protein, fetal and adult testis expressed 1, fetal and adult testis-expressed transcript protein, Tumor antigen BJ-HCC-2
Mouse
Affinity Purified
Primary
Human
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.