Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FATE1 Antibody (OTI1A1) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272310
Description
FATE1 Monoclonal specifically detects FATE1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
FATE1 | |
Monoclonal | |
LYOPH | |
Western Blot 1:2000, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence 1:100 | |
Cancer/testis antigen 43, CT43BJ-HCC-2 antigen, FATEfetal and adult testis expressed transcript protein, fetal and adult testis expressed 1, fetal and adult testis-expressed transcript protein, Tumor antigen BJ-HCC-2 | |
Mouse | |
20.5 kDa | |
100 μg | |
89885 | |
Human | |
Lyophilized |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI1A1 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
FATE1 | |
Full length human recombinant protein of human FATE1(NP_149076) produced in HEK293T cell. | |
Affinity Purified | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction