Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FATE1 Antibody (OTI1A1), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272310R
Description
FATE1 Monoclonal specifically detects FATE1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
FATE1 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
FATE1 | |
Full length recombinant protein of human FATE1 (NP_149076) produced in HEK293T cells. | |
0.1 mL | |
89885.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI1A1 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
Cancer/testis antigen 43, CT43BJ-HCC-2 antigen, FATEfetal and adult testis expressed transcript protein, fetal and adult testis expressed 1, fetal and adult testis-expressed transcript protein, Tumor antigen BJ-HCC-2 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction