Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FATE1 Monoclonal Antibody (OTI6A11)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA526085
Description
FATE1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes a cancer-testis antigen that is highly expressed in hepatocellular carcinomas and other tumors and weakly expressed in normal tissues except testis. The protein is strongly expressed in spermatogonia, primary spermatocytes, and Sertoli cells in seminiferous tubules. This protein may have a role in the control of early testicular differentiation and cell proliferation.
Specifications
FATE1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q969F0 | |
FATE1 | |
Full length human recombinant protein of FATE1 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
OTI6A11 | |
Unconjugated | |
FATE1 | |
9230118O15Rik; BJ-HCC-2 antigen; cancer/testis antigen 43; CT43; EGK_21049; FATE; FATE1; fetal and adult testis expressed 1; fetal and adult testis expressed transcript protein; fetal and adult testis-expressed transcript protein; Tumor antigen BJ-HCC-2 | |
Mouse | |
Affinity chromatography | |
RUO | |
89885 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction