Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FATE1 Monoclonal Antibody (OTI6G10), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA505265S


Description
FATE1 Monoclonal antibody specifically detects FATE1 in Canine, Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
FATE1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P60604, P60605 | |
Mouse | |
Affinity chromatography | |
RUO | |
22213, 294331, 611581, 7327 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Western Blot | |
OTI6G10 | |
Unconjugated | |
UBE2G2 | |
9230118O15Rik; BJ-HCC-2 antigen; cancer/testis antigen 43; CT43; EGK_21049; FATE; FATE1; fetal and adult testis expressed 1; fetal and adult testis expressed transcript protein; fetal and adult testis-expressed transcript protein; Tumor antigen BJ-HCC-2 | |
Full length human recombinant protein of human UBE2G2 produced in HEK293T cell. | |
30 μL | |
Primary | |
Canine, Human, Mouse, Rat | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction