Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FATE1 Monoclonal Antibody (OTI7B5), TrueMAB™, OriGene
SDP

Catalog No. 502539739
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-253-9739 30 μL
50-167-5868 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-253-9739 Supplier OriGene Supplier No. TA505383S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FATE1 Monoclonal antibody specifically detects FATE1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen FATE1
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI7B5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Egr2
Gene Accession No. P11161
Gene Alias 9230118O15Rik; BJ-HCC-2 antigen; cancer/testis antigen 43; CT43; EGK_21049; FATE; FATE1; fetal and adult testis expressed 1; fetal and adult testis expressed transcript protein; fetal and adult testis-expressed transcript protein; Tumor antigen BJ-HCC-2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 156-476 of human EGR2 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1959
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.