Learn More
FATE1 Mouse anti-Human, Clone: OTI5E2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505183

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a cancer-testis antigen that is highly expressed in hepatocellular carcinomas and other tumors and weakly expressed in normal tissues except testis. The protein is strongly expressed in spermatogonia, primary spermatocytes, and Sertoli cells in seminiferous tubules. This protein may have a role in the control of early testicular differentiation and cell proliferation.Specifications
FATE1 | |
Monoclonal | |
Unconjugated | |
FATE1 | |
CT43, FATE | |
Mouse | |
Affinity Chromatography | |
RUO | |
89885 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI5E2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q969F0 | |
FATE1 | |
Full length human recombit protein of human FATE1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.