Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Fatostatin A
GREENER_CHOICESDP

Catalog No. 444450 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-445-0 50 mg
44-441-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 44-445-0 Supplier R&D Systems™ Supplier No. 4444/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Cell permeable inhibitor of SREBP activation

Inhibitor of sterol regulatory element binding protein (SREBP); impairs the activation of SREBP-1 and SREBP-2. Exhibits antiproliferative effects in DU 145 cells independently of IGF-1 signaling (IC50 = 0.1 μM); reverses hyperglycemia in diabetic (ob/ob) mice. Cell permeable.

Chemical Identifiers

CAS 298197-04-3
Molecular Formula C18H19BrN2S

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 375.33
Purity >97%
Molecular Weight (g/mol) 375.328
Chemical Name or Material Fatostatin A
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.