Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FATP1/SLC27A1 Antibody (308420) [DyLight 680], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB3304Y
Description
FATP1/SLC27A1 Monoclonal antibody specifically detects FATP1/SLC27A1 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOFSpecifications
FATP1/SLC27A1 | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
376497 | |
Store at 4°C in the dark. | |
IgG2b |
Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
308420 | |
Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
ACSVL5FATP-1, EC 6.2.1, EC 6.2.1.-, FATP1Solute carrier family 27 member 1, FATPlong-chain fatty acid transport protein 1, Fatty acid transport protein 1, FLJ00336, MGC71751, solute carrier family 27 (fatty acid transporter), member 1 | |
NS0 mouse myeloma cell line transfected with human FATP1, Met1-Leu646, Accession # Q6PCB7 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction