Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FATP2/SLC27A2 Antibody (6B3A9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$560.50
Specifications
Antigen | FATP2/SLC27A2 |
---|---|
Clone | 6B3A9 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Description
FATP2/SLC27A2 Monoclonal specifically detects FATP2/SLC27A2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
FATP2/SLC27A2 | |
1 mg/mL | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS with 0.05% Sodium Azide | |
11001 | |
Purified recombinant fragment of human SLC27A2 (AA: 346-405) expressed in E. Coli. | |
Primary | |
70.3 kDa |
6B3A9 | |
Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
Monoclonal | |
Mouse | |
Human | |
ACSVL1FATP-2, EC 6.2.1, EC 6.2.1.-, EC 6.2.1.3, EC 6.2.1.7, FACVL1very long-chain fatty-acid-coenzyme A ligase 1, FATP2THCA-CoA ligase, Fatty acid transport protein 2, Fatty-acid-coenzyme A ligase, very long-chain 1, hFACVL1, HsT17226, Long-chain-fatty-acid--CoA ligase, solute carrier family 27 (fatty acid transporter), member 2, Solute carrier family 27 member 2, very-long-chain acyl-CoA synthetase, VLACSVery long-chain-fatty-acid-CoA ligase, VLCSvery long-chain acyl-CoA synthetase | |
SLC27A2 | |
IgG1 | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title