Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FATP4/SLC27A4 Antibody (342142), PE/Cy5.5, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB3650PECY55
Description
FATP4/SLC27A4 Monoclonal antibody specifically detects FATP4/SLC27A4 in Human samples. It is validated for Flow CytometrySpecifications
FATP4/SLC27A4 | |
Monoclonal | |
PE-Cyanine5.5 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
10999 | |
Store at 4C in the dark. Do not freeze. | |
IgG2b |
Flow Cytometry | |
342142 | |
Flow Cytometry | |
ACSVL4Solute carrier family 27 member 4, EC 6.2.1, EC 6.2.1.-, EC 6.2.1.7, FATP-4, FATP4Fatty acid transport protein 4, IPS, long-chain fatty acid transport protein 4, solute carrier family 27 (fatty acid transporter), member 4 | |
NS0 mouse myeloma cell line transfected with human FATP4, Met1-Leu643, Accession # Q6P1M0 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction