Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fatty acid desaturase 2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | Fatty acid desaturase 2 |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Fatty acid desaturase 2 Polyclonal specifically detects Fatty acid desaturase 2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Fatty acid desaturase 2 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
9415 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:GKGGNQGEGAAEREVSVPTFSWEEIQKHNLRTDRWLVIDRKVYNITKWSIQHPGGQRVIGHYAGEDATDAFRAFHPDLEFVGKFLKPLLIGELAPEEPSQDHGKNSKITEDFRALRKTAEDMNLFKTNH | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Polyclonal | |
Rabbit | |
Human | |
D6DLLCDL2, Delta(6) desaturase, Delta(6) fatty acid desaturase, Delta-6 desaturase, delta-6 fatty acid desaturase, delta-6-desaturase, DES6, EC 1.14.19, EC 1.14.19.-, FADSD6, fatty acid desaturase 2, linoleoyl-CoA desaturase (delta-6-desaturase)-like 2, SLL0262, TU13 | |
FADS2 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title