Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fatty Acid Ethyl Esters (FAEES), C4 - C24 Even Carbon Saturated, Certified Reference Material, 1,000 μg/mL, MilliporeSigma™ Supelco™
Click to view available options
Quantity:
1 mL
Description
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications
Specifications
Packaging | Ampule |
Product Line | TraceCERT |
Physical Form | Liquid |
Concentration | 1000 μg/mL each component in hexane |
Quantity | 1 mL |
Grade | TraceCERT™ Analytical Standard |
Concentration or Composition (by Analyte or Components) | Ethyl butyrate; Ethyl hexanoate; Ethyl octanoate; Ethyl decanoate; Ethyl dodecanoate; Ethyl myristate; Ethyl stearate; Ethyl behenate; Ethyl arachidate; Ethyl palmitate; Ethyl tetracosanoate |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction