Learn More
Description
Specifications
Specifications
| Antigen | Fatty Acid Synthase/FASN |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence 1:2000, Immunoprecipitation 1:100, Immunohistochemistry-Paraffin 1:500immunoprecipitation 1:100, Knockdown Validated |
| Formulation | Whole antisera with 0.05% Sodium Azide |
| Gene Accession No. | P19096 |
| Gene Alias | EC 2.3.1, EC 2.3.1.85, EC 6.3.3.1, FASshort chain dehydrogenase/reductase family 27X, member 1, fatty acid synthase, MGC14367, MGC15706, OA-519, SDR27X1 |
| Gene Symbols | FASN |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
