Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAU Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100519
Description
Antibody detects endogenous levels of total FAU.
This gene is the cellular homolog of the fox sequence in the Finkel-Biskis-Reilly murine sarcoma virus (FBR-MuSV). It encodes a fusion protein consisting of the ubiquitin-like protein fubi at the N terminus and ribosomal protein S30 at the C terminus. It has been proposed that the fusion protein is post-translationally processed to generate free fubi and free ribosomal protein S30. Fubi is a member of the ubiquitin family, and ribosomal protein S30 belongs to the S30E family of ribosomal proteins. Whereas the function of fubi is currently unknown, ribosomal protein S30 is a component of the 40S subunit of the cytoplasmic ribosome and displays antimicrobial activity. Pseudogenes derived from this gene are present in the genome. Similar to ribosomal protein S30, ribosomal proteins S27a and L40 are synthesized as fusion proteins with ubiquitin.
Specifications
FAU | |
Polyclonal | |
Unconjugated | |
FAU | |
40S ribosomal protein S30; 40S ribosomal protein S30; ubiquitin-like protein fubi and ribosomal protein S30; asr1; Fau; FAU ubiquitin like and ribosomal protein S30 fusion; FAU, ubiquitin like and ribosomal protein S30 fusion; FAU1; FAU-encoded ubiquitin-like protein; FBR-MuSV-associated ubiquitously expressed; Finkel-Biskis-Reilly murine sarcoma virus (FBR-MuSV) ubiquitously expressed; Finkel-Biskis-Reilly murine sarcoma virus (FBR-MuSV) ubiquitously expressed (fox derived); Finkel-Biskis-Reilly murine sarcoma virus (FBR-MuSV) ubiquitously expressed (fox derived) protein; Finkel-Biskis-Reilly murine sarcoma virus (FBR-MuSV) ubiquitously expressed (fox derived); ribosomal protein S30; Finkel-Biskis-Reilly murine sarcoma virusubiquitously expressed; Fub1; Fubi; MNSFbeta; MNSF-beta; monoclonal nonspecific suppressor factor beta; Monoclonal non-specific suppressor factor beta; monoclonal nonspecific suppressor factor betamonoclonal nonspecific suppressor factor beta; ribosomal protein S30; RP MNSFbeta; RPS30; S30; ubiquitin-like protein FUBI; ubiquitin-like protein fubi and ribosomal protein S30; ubiquitin-like/S30 ribosomal fusion protein; ubiquitin-like-S30 fusion protein | |
Rabbit | |
Affinity chromatography | |
RUO | |
14109, 2197, 29752 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P62861, P62862, P62864 | |
FAU | |
A synthesized peptide derived from human FAU(Accession P62861), corresponding to amino acid residues A9-K28. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction