Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAU Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111476
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111476 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111476 Supplier Invitrogen™ Supplier No. PA5111476
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KTGRAKRRMQ YNRRFVNVVP TFGKKKGPNA N Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.

This gene is the cellular homolog of the fox sequence in the Finkel-Biskis-Reilly murine sarcoma virus (FBR-MuSV). It encodes a fusion protein consisting of the ubiquitin-like protein fubi at the N terminus and ribosomal protein S30 at the C terminus. It has been proposed that the fusion protein is post-translationally processed to generate free fubi and free ribosomal protein S30. Fubi is a member of the ubiquitin family, and ribosomal protein S30 belongs to the S30E family of ribosomal proteins. Whereas the function of fubi is currently unknown, ribosomal protein S30 is a component of the 40S subunit of the cytoplasmic ribosome and displays antimicrobial activity. Pseudogenes derived from this gene are present in the genome. Similar to ribosomal protein S30, ribosomal proteins S27a and L40 are synthesized as fusion proteins with ubiquitin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAU
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAU
Gene Accession No. P62861
Gene Alias 40S ribosomal protein S30; 40S ribosomal protein S30; ubiquitin-like protein fubi and ribosomal protein S30; asr1; Fau; FAU ubiquitin like and ribosomal protein S30 fusion; FAU, ubiquitin like and ribosomal protein S30 fusion; FAU1; FAU-encoded ubiquitin-like protein; FBR-MuSV-associated ubiquitously expressed; Finkel-Biskis-Reilly murine sarcoma virus (FBR-MuSV) ubiquitously expressed; Finkel-Biskis-Reilly murine sarcoma virus (FBR-MuSV) ubiquitously expressed (fox derived); Finkel-Biskis-Reilly murine sarcoma virus (FBR-MuSV) ubiquitously expressed (fox derived) protein; Finkel-Biskis-Reilly murine sarcoma virus (FBR-MuSV) ubiquitously expressed (fox derived); ribosomal protein S30; Finkel-Biskis-Reilly murine sarcoma virusubiquitously expressed; Fub1; Fubi; MNSFbeta; MNSF-beta; monoclonal nonspecific suppressor factor beta; Monoclonal non-specific suppressor factor beta; monoclonal nonspecific suppressor factor betamonoclonal nonspecific suppressor factor beta; ribosomal protein S30; RP MNSFbeta; RPS30; S30; ubiquitin-like protein FUBI; ubiquitin-like protein fubi and ribosomal protein S30; ubiquitin-like/S30 ribosomal fusion protein; ubiquitin-like-S30 fusion protein
Gene Symbols FAU
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human FAU. Recombinant protein control fragment (Product #RP-101930).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2197
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.