Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Favipiravir
GREENER_CHOICESDP

Catalog No. 722550 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-255-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 72-255-0 Supplier Tocris Bioscience™ Supplier No. 7225/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Viral RNA polymerase inhibitor

Favipiravir is a viral RNA polymerase inhibitor; exhibits no effects on mammalian RNA polymerase. Prodrug that is phosphoribosylated by cellular enzymes to its active form, Favipiravir-ribofuranosyl-5'-triphosphate (RTP). Reduces infection rate of SARS-CoV-2 in Vero E6 cells in vitro (IC50 = 61.88 μM). Inhibits influenza virus RNA-dependent RNA polymerase (RdRP) activity (IC50 = 341 nM). Induces lethal mutagenesis in MDCK cells inoculated with seasonal influenza A (H1N1) viruses. Protects mice against Ebola virus.

Specifications

Chemical Name or Material 6-Fluoro-3,4-dihydro-3-oxo-2-pyrazinecarboxamide
CAS 259793-96-9
Quantity 50 mg
Target RNA/DNA Polymerase Inhibitors
Molecular Formula C5H4FN3O2
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.