Learn More
Fisherbrand™ Grey Safety Goggles
Catalog No. 19181513 Shop All Fisherbrand Products
Change view
Click to view available options
Product Type:
Safety/Chemical Splash Goggles
Catalog No. Product Type
19-181-513 Safety/Chemical Splash Goggles
1 options
Catalog No. 19-181-513
Only null left
Add to Cart
Edge
Add to Cart
Polyester headband provides a comfortable fit, while indirect venting allows air flow for anti-fog protection.
Streamlined design to work with hardhats and respirators.
  • Polycarbonate lens with anti-fog and anti-scratch coating
  • Frame consists of a combination of a polycarbonate lens with a Thermoplastic rubber lining
  • Provides chemical splash protection
  • Indirect venting allows airflow to minimize fogging
  • Comfortable polyester headband provides a comfortable fit
  • ANSI/ISEA Z87.1-2020: Z87+ U6 D3 D4
Certifications
ANSI Z87
Compliance
Meets ANSI/ISEA Z87.1-2020: Z87+ U6 D3 D4 standards.
Specifications
Certifications/Compliance ANSI/ISEA Z87.1-2020: Z87+ U6 D3 D4
Lens Coating Anti-fog, Scratch-resistant
Lens Tint Clear
Product Type Safety/Chemical Splash Goggles
Frame Color Light Gray
Material Polyester
Frame Material Polycarbonate, TPR