Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBG3 (F Box 3) Polyclonal Antibody, QED Bioscience™
SDP

Catalog No. 502277925
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7925 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-227-7925 Supplier Leinco Technologies Inc Supplier No. 28104100UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FBG3 (F Box 3) Polyclonal antibody specifically detects FBG3 (F Box 3) in Human samples. It is validated for Western Blot

Specifications

Antigen FBG3 (F Box 3)
Applications Western Blot
Classification Polyclonal
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene FBXO44
Gene Accession No. Q9H4M3
Gene Alias 5730411K09; AV001623; DKFZp781J0852; F Box Gama 3; FBG3; F-box gene 3; F-box only protein 44; F-box only protein 6a; F-box protein 44; F-box protein FBX30; F-box/G-domain protein 3; FBX30; Fbx44; FBX6A; FBXO44; FBXO6A; MGC14140; RGD1562463; RP3-330O12.2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 103-119 (EWKVEDLSRDQRKEFPN) of human
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93611
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.