Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FBG4 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100466MFV610
Description
FBG4 Polyclonal antibody specifically detects FBG4 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| FBG4 | |
| Polyclonal | |
| Western Blot | |
| FBG4FBXO26, F-box only protein 17, F-box protein 17, F-box protein FBG4, Fbx17, FBX26F-box only protein 26, FLJ11798, FLJ25205, FLJ58300, MGC9379 | |
| A synthetic peptide contianing residues 70-85 (LQLARDRSAEGRALYA) of the human FBG4 protein. There is 100% sequence identity between the immunogen and mouse and rat sequences. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 115290 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction