Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBG4 (F Box 4) Polyclonal Antibody, QED Bioscience™
SDP

Catalog No. 502277926
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7926 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-7926 Supplier Leinco Technologies Inc Supplier No. 28105100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FBG4 (F Box 4) Polyclonal antibody specifically detects FBG4 (F Box 4) in Human samples. It is validated for Western Blot

Specifications

Antigen FBG4 (F Box 4)
Applications Western Blot
Classification Polyclonal
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene FBXO17
Gene Accession No. Q96EF6
Gene Alias F Box Gama 4; FBG4; F-box only protein 17; F-box only protein 26; F-box protein 17; F-box protein FBG4; FBX17; FBX26; Fbxo17; FBXO26; FLJ11798; FLJ25205; FLJ58300; MGC9379
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 70-85 (LQLARDRSAEGRALYA) of human FBG4 conjugated to KLH at the N-terminus.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115290
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.