Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBG4 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA123512

Catalog No. PIPA123512


Only null left
Add to cart

Description

Description

PA1-23512 has been successfully used in ELISA and Western Blot procedures.

F-box proteins, a critical component of the evolutionary conserved ubiquitin-protein ligase complex SCF (Skp1/Cdc53-Cullin1/F-box), recruit substrates for ubiquitination and consequent degradation through their specific protein-protein interaction domains. Novel human F-box proteins named FBG1, FBG2, FBG3, FBG4 and FBG5 constitute a third subfamily of mammalian F-box proteins. FBG genes are expressed in a limited number of human tissues including kidney, liver, brain and muscle tissues.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FBG4
Polyclonal
Unconjugated
FBXO17
F Box Gama 4; FBG4; F-box only protein 17; F-box only protein 26; F-box protein 17; F-box protein FBG4; FBX17; FBX26; Fbxo17; FBXO26; FLJ11798; FLJ25205; FLJ58300; MGC9379
Rabbit
Affinity chromatography
RUO
115290
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q96EF6
FBXO17
Rabbit Polyclonal to FBG4 ( F Box 4 ).
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.