Learn More
Invitrogen™ FBG4 Polyclonal Antibody

Description
PA1-4349 detects FBG4 from human, mouse and rat samples. PA1-4349 has been successfully used in Western blot procedures. By Western blot this antibody detects a ~31 kDa protein representing FBG4. PA1-4349 immunizing peptide corresponds to amino acid residues 70-85 from human FBG4 protein. Maintain antibody at 4ºC for up to 3 months. Store at -20ºC long term, avoiding freeze/thaw cycles.
Specifications
Specifications
| Antigen | FBG4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | FBXO17 |
| Gene Accession No. | Q6AY27, Q96EF6, Q9QZM8 |
| Gene Alias | F Box Gama 4; FBG4; F-box only protein 17; F-box only protein 26; F-box protein 17; F-box protein FBG4; FBX17; FBX26; Fbxo17; FBXO26; FLJ11798; FLJ25205; FLJ58300; MGC9379 |
| Gene Symbols | FBXO17 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.