Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBG4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114264
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114264 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114264 Supplier Invitrogen™ Supplier No. PA5114264
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VQVLSHVPPR SLVTRCRPVC RAWRDIVDGP TVWLLQLARD RSAEGRALYA For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 92%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 86%; Rat: 100%.

F-box proteins, a critical component of the evolutionary conserved ubiquitin-protein ligase complex SCF (Skp1/Cdc53-Cullin1/F-box), recruit substrates for ubiquitination and consequent degradation through their specific protein-protein interaction domains. Novel human F-box proteins named FBG1, FBG2, FBG3, FBG4 and FBG5 constitute a third subfamily of mammalian F-box proteins. FBG genes are expressed in a limited number of human tissues including kidney, liver, brain and muscle tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBG4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FBXO17
Gene Accession No. Q96EF6
Gene Alias F Box Gama 4; FBG4; F-box only protein 17; F-box only protein 26; F-box protein 17; F-box protein FBG4; FBX17; FBX26; Fbxo17; FBXO26; FLJ11798; FLJ25205; FLJ58300; MGC9379
Gene Symbols FBXO17
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal region of Human FBXO17 (aa 27-76).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115290
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.