Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fbl14 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP17952020UL
Description
Fbl14 Polyclonal specifically detects Fbl14 in Human samples. It is validated for Western Blot.Specifications
| Fbl14 | |
| Polyclonal | |
| Western Blot 1:1000 | |
| NP_689654 | |
| FBXL14 | |
| Synthetic peptide directed towards the N terminal of human FBXL14. Peptide sequence WRDAAYHKSVWRGVEAKLHLRRANPSLFPSLQARGIRRVQILSLRRSLSY. | |
| 20 μL | |
| Primary | |
| Human | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS and 2% Sucrose with 0.09% Sodium Azide | |
| FBL14, F-box and leucine-rich repeat protein 14 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 144699 | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction