Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FBLIM1 Monoclonal Antibody (OTI3G1)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA526856
Description
FBLIM1 Monoclonal Antibody for Western Blot
This gene encodes a protein with an N-terminal filamin-binding domain, a central proline-rich domain, and, multiple C-terminal LIM domains. This protein localizes at cell junctions and may link cell adhesion structures to the actin cytoskeleton. This protein may be involved in the assembly and stabilization of actin-filaments and likely plays a role in modulating cell adhesion, cell morphology and cell motility. This protein also localizes to the nucleus and may affect cardiomyocyte differentiation after binding with the CSX/NKX2-5 transcription factor. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
| FBLIM1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8WUP2 | |
| FBLIM1 | |
| Human recombinant protein fragment corresponding to amino acids 1-242 of FBLIM1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI3G1 | |
| Unconjugated | |
| FBLIM1 | |
| 2410043F08Rik; CAL; CSX-associated LIM; FBLIM1; FBLP1; FBLP-1; filamin binding LIM protein 1; filamin-binding LIM protein 1; filamin-binding LIM protein-1; Gt10; MIG2-interacting protein; migfilin; migfilin(s); mitogen-inducible 2 interacting protein; mitogen-inducible 2-interacting protein | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 54751 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction