Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBLL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549014
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549014 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549014 Supplier Invitrogen™ Supplier No. PA549014
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with mouse based on sequence homology.

S-adenosyl-L-methionine-dependent methyltransferase that has the ability to methylate both RNAs and proteins. Involved in pre-rRNA processing by catalyzing the site-specific 2'-hydroxyl methylation of ribose moieties in pre-ribosomal RNA. Also acts as a protein methyltransferase by mediating methylation of glutamine residues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBLL1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene FBLL1
Gene Accession No. A6NHQ2
Gene Alias FBLL1; fibrillarin-like 1
Gene Symbols FBLL1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 209-236 amino acids from the Central region of human FBLL1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 345630
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.