Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBP1/FBP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552943
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552943 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552943 Supplier Invitrogen™ Supplier No. PA552943
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RKAGLAHLYG IAGSVNVTGD EVKKLDVLSN SLVINMVQSS YSTCVLVSEE NKDAIITAKE KRGKYVVCFD PLDG This product detects both FBP1 and FBP2, which are 80% homologous. The antigen sequence is a 100% match to FBP2 and the product has been validated against FBP1. Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 93%.

FBP1 and FBP2 are gluconeogenesis regulatory enzymes which catalyze the hydrolosis of fructose 1,6-bisphosphate to fructose 6-phosphate and inorganic phosphate. Fructose-1,6-diphosphatase deficiency is associated with hypoglycemia and metabolic acidosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBP1/FBP2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FBP1
Gene Accession No. O00757, P09467
Gene Alias D-fructose-1,6-bisphosphate 1-phosphohydrolase 1; D-fructose-1,6-bisphosphate 1-phosphohydrolase 2; FBP; FBP1; FBP2; FBPase 1; FBPase 2; fructose-1; Fructose-1,6-bisphosphatase 1; fructose-1,6-bisphosphatase 2; fructose-1,6-bisphosphatase isozyme 2; fructose-bisphosphatase 1; fructose-bisphosphatase 2; growth-inhibiting protein 17; hexosediphosphatase; liver FBPase; Muscle FBPase; muscle fructose-bisphosphatase
Gene Symbols FBP1, FBP2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FBP2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2203, 8789
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.