Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBP1 Recombinant Rabbit Monoclonal Antibody (7N8C1)
GREENER_CHOICE

Catalog No. PIMA535149
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535149 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535149 Supplier Invitrogen™ Supplier No. MA535149
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: APYGARYVGS MVADVHRTLV YGGIFLYPAN KKSPNGKLRL LYECNPMAYV MEKAGGMATT GKEAVLDVIP TDIHQRAPVI LGSPDDVLEF LKVYEKHSAQ.

Fructose-1,6-bisphosphatase 1, a gluconeogenesis regulatory enzyme, catalyzes the hydrolysis of fructose 1,6-bisphosphate to fructose 6-phosphate and inorganic phosphate. Fructose-1,6-diphosphatase deficiency is associated with hypoglycemia and metabolic acidosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBP1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 7N8C1
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FBP1
Gene Accession No. P09467, P19112, Q9QXD6
Gene Alias D-fructose-1,6-bisphosphate 1-phosphohydrolase 1; D-fructose-1,6-bisphosphate 1-phosphohydrolase 3; EC 3.1.3.11; Fbp; FBP 1; FBP1; Fbp2; Fbp-2; Fbp3; FBPase 1; FBPase 3; FBPase brain isoform; FBPase liver; Fdp; fructose bisphosphatase 1; fructose-1; fructose-1,6- biphosphatase 1; fructose-1,6-bisphosphatase; fructose-1,6-bisphosphatase 1; fructose-1,6-bisphosphatase 2; fructose-1,6-bisphosphatase isozyme 3; Fructose-16- biphosphatase; fructose-bisphosphatase 1; growth-inhibiting protein 17; liver FBPase
Gene Symbols FBP1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 239-338 of human FBP1 (P09467).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14121, 2203, 24362
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.