Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582222
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582222 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582222 Supplier Invitrogen™ Supplier No. PA582222
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DCLVSVGTIF GIYRKKSTDE PSEKDALQPG RNLVAAGYAL YGSATMLVLA MDCGVNCFML DPAIGEFILV DKDVKIKKKG KIYSLNEGYA KDFDPAVTEY IQRKKFPPDN SAP Highest antigen sequence indentity to the following orthologs: Rat - 88%, Mouse - 90%.

Fructose-1,6-bisphosphatase 1, a gluconeogenesis regulatory enzyme, catalyzes the hydrolysis of fructose 1,6-bisphosphate to fructose 6-phosphate and inorganic phosphate. Fructose-1,6-diphosphatase deficiency is associated with hypoglycemia and metabolic acidosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FBP1
Gene Accession No. P09467
Gene Alias D-fructose-1,6-bisphosphate 1-phosphohydrolase 1; D-fructose-1,6-bisphosphate 1-phosphohydrolase 3; EC 3.1.3.11; Fbp; FBP 1; FBP1; Fbp2; Fbp-2; Fbp3; FBPase 1; FBPase 3; FBPase brain isoform; FBPase liver; Fdp; fructose bisphosphatase 1; fructose-1; fructose-1,6- biphosphatase 1; fructose-1,6-bisphosphatase; fructose-1,6-bisphosphatase 1; fructose-1,6-bisphosphatase 2; fructose-1,6-bisphosphatase isozyme 3; Fructose-16- biphosphatase; fructose-bisphosphatase 1; growth-inhibiting protein 17; liver FBPase
Gene Symbols FBP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FBP1. Recombinant protein control fragment (Product #RP-88835).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2203
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.