Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109199
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109199 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109199 Supplier Invitrogen™ Supplier No. PA5109199
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: Mouse liver tissue, rat liver tissue, rat bladder tissue, human liver tissue, human thyroid carcinoma tissue, human kidney tissue, mouse pancreas tissue.

Fructose-1,6-bisphosphatase 1, a gluconeogenesis regulatory enzyme, catalyzes the hydrolysis of fructose 1,6-bisphosphate to fructose 6-phosphate and inorganic phosphate. Fructose-1,6-diphosphatase deficiency is associated with hypoglycemia and metabolic acidosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 40% glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4
Gene FBP1
Gene Accession No. P09467
Gene Alias D-fructose-1,6-bisphosphate 1-phosphohydrolase 1; D-fructose-1,6-bisphosphate 1-phosphohydrolase 3; EC 3.1.3.11; Fbp; FBP 1; FBP1; Fbp2; Fbp-2; Fbp3; FBPase 1; FBPase 3; FBPase brain isoform; FBPase liver; Fdp; fructose bisphosphatase 1; fructose-1; fructose-1,6- biphosphatase 1; fructose-1,6-bisphosphatase; fructose-1,6-bisphosphatase 1; fructose-1,6-bisphosphatase 2; fructose-1,6-bisphosphatase isozyme 3; Fructose-16- biphosphatase; fructose-bisphosphatase 1; growth-inhibiting protein 17; liver FBPase
Gene Symbols FBP1
Host Species Rabbit
Immunogen Recombinant protein within Human FBP1 aa 1-140.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2203
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.