Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FBP1 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS3980R
Description
Fructose-1,6-bisphosphatase 1, a gluconeogenesis regulatory enzyme, catalyzes the hydrolysis of fructose 1,6-bisphosphate to fructose 6-phosphate and inorganic phosphate. Fructose-1,6-diphosphatase deficiency is associated with hypoglycemia and metabolic acidosis.Specifications
| FBP1 | |
| Polyclonal | |
| Unconjugated | |
| FBP1 | |
| BOS_9181; D-fructose-1,6-bisphosphate 1-phosphohydrolase 1; D-fructose-1,6-bisphosphate 1-phosphohydrolase 3; EC 3.1.3.11; Fbp; FBP 1; FBP1; Fbp2; Fbp-2; Fbp3; FBPase 1; FBPase 3; FBPase brain isoform; FBPase liver; Fdp; fructose bisphosphatase 1; fructose-1,6- biphosphatase 1; Fructose-1,6-bisphosphatase 1; Fructose-1,6-bisphosphatase isozyme 3; Fructose-16- biphosphatase; fructose-bisphosphatase 1; growth-inhibiting protein 17; Liver FBPase | |
| Rabbit | |
| Protein A | |
| RUO | |
| 2203 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1 μg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
| P09467 | |
| FBP1 | |
| KLH conjugated synthetic peptide derived from human FBP1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction