Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FBP11 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | FBP11 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-8532
![]() |
Bethyl Laboratories, Inc
A304736AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
FNBP3, Formin-binding protein 3, is a 957 amino acid protein encoded by the human gene PRPF40A. FNBP3 belongs to the PRPF40 family and contains five FF domains and two WW domains. Nuclear proteins harboring both WW and FF protein interaction modules bind to splicing factors as well as RNA polymerase II and may serve to link transcription with splicing. Through the WW domains, FNBP3 will bind to the Formin proline-rich regions. Also known as Pre-mRNA-processing factor 40 homolog A, FNBP3 binds to WASL/N-WASP (Neuronal Wiskott-Aldrich syndrome protein) complex and suppresses its translocation from the nucleus to the cytoplasm, thereby inhibiting its cytoplasmic function. FNBP3 is widely expressed in most tissues and is localized to the nuclear speckles.Specifications
FBP11 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
fas ligand-associated factor 1, Fas-ligand associated factor 1, FBP11, FBP-11, FLAF1, FLJ20585, FNBP3, formin binding protein 3, formin-binding protein 11, Formin-binding protein 3, HIP10, HIP-10, huntingtin yeast partner A, huntingtin-interacting protein 10, Huntingtin-interacting protein A, HYPA, NY-REN-6, NY-REN-6 antigen, Prp40, PRP40 pre-mRNA processing factor 40 homolog A, PRP40 pre-mRNA processing factor 40 homolog A (S. cerevisiae), PRP40 pre-mRNA processing factor 40 homolog A (yeast), renal carcinoma antigen NY-REN-6 | |
PRPF40A | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
O75400 | |
55660 | |
Between 275 and 325 | |
Primary | |
4° C | |
PRPF40A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title