Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human FBP2 His Protein
SDP

Catalog No. p-7105134
Click to view available options
Quantity:
0.02 mg
0.1 mg

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-339 of Human FBP2 The Recombinant Human FBP2 Protein is derived from E. coli. The Recombinant Human FBP2 Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.

Specifications

Concentration 1mg/mL
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) containing 0.15M NaCl, 10% glycerol, 1mM DTT
Gene ID (Entrez) 8789
Molecular Weight (g/mol) TMW: 39kDa
Quantity 0.02 mg
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias D-fructose-1,6-bisphosphate 1-phosphohydrolase 2, EC 3.1.3, EC 3.1.3.11, FBPase 2, fructose-1,6-bisphosphatase 2, fructose-1,6-bisphosphatase isozyme 2, hexosediphosphatase, MGC142192, muscle fructose-bisphosphatase
Common Name FBP2
Biological Activity Specific activity is > 1,500 pmol/min/ug obtained bymeasuring the increase of NADPH absorbance at 340 nm resulting from thereduction of NADP. One unit will oxidize 1.0 pmole of fructose 1,6 diphosphate to fructose 6-phosphate and inorganic phosphate per minute at pH 9.5 at 37C.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Show More Show Less

For research use only and is not approved for use in humans or in clinical diagnosis

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.