Learn More
Invitrogen™ FBP2 Polyclonal Antibody

Description
Immunogen sequence: VAYIIEQAGGL ATTGTQPVLD VKPEAIHQRV PLILGSPEDV QEYLTCVQKN QAGS Highest antigen sequence identity to the following orthologs - mouse 91%, rat 91%.
Specifications
Specifications
| Antigen | FBP2 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | FBP2 |
| Gene Accession No. | O00757 |
| Gene Alias | D-fructose-1,6-bisphosphate 1-phosphohydrolase 2; Fbp1; Fbp-1; FBP2; FBPase; FBPase 2; FBPase muscle; fructose bisphosphatase 2; fructose-1; fructose-1,6-bisphosphatase 2; fructose-1,6-bisphosphatase isozyme 2; fructose-bisphosphatase 2; hexosediphosphatase; muscle FBPase; muscle fructose-bisphosphatase; Rae-30 |
| Gene Symbols | FBP2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.