Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FBPase 1 Antibody (4G3), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | FBPase 1 |
---|---|
Clone | 4G3 |
Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
Applications | ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
FBPase 1 Monoclonal antibody specifically detects FBPase 1 in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
FBPase 1 | |
ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
D-fructose-1,6-bisphosphate 1-phosphohydrolase 1, EC 3.1.3, EC 3.1.3.11, FBP, FBPase 1, fructose-1,6-bisphosphatase 1, fructose-bisphosphatase 1, growth-inhibiting protein 17 | |
A synthesized peptide derived from Human FBPase 1 [UniProt P09467] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
4G3 | |
ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Lipid and Metabolism, Protein Phosphatase | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
2203 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title