Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBRS Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5122002
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5122002 20 μL
PIPA5122001 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5122002 Supplier Invitrogen™ Supplier No. PA5122002
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Fibrosin is a lymphokine secreted by activated lymphocytes that induces fibroblast proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBRS
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene Fbrs
Gene Accession No. Q8R089, Q9HAH7
Gene Alias Fbrs; FBS; FBS1; fbsh; fibrogenic lymphokine; fibrosin; fibrosin 1; fibrosin-1; FLJ11618; probable fibrosin-1; probable fibrosin-1 long transcript protein
Gene Symbols Fbrs
Host Species Rabbit
Immunogen Met1-Asp250 (Accession Q9HAH7), with N-terminal His Tag.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14123, 64319
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.