Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBW7 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501571941
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-1941 100 μL
50-157-1942 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-1941 Supplier Bethyl Laboratories, Inc Supplier No. IHC00348
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Monkey, Gorilla, Chimpanzee, Northern white-cheeked gibbon and Crab-eating macaque

Cyclin E, one of the activators of the cyclin-dependent kinase Cdk2, is critical for the initiation of DNA replication and other S-phase functions. The proper timing and amplitude of cyclin E expression seem to be important, because elevated levels of cyclin E have been associated with a variety of maligcies and constitutive expression of cyclin E leads to genomic instability. Phosphorylated cyclin E depends on the novel human gene, hCdc4, that was found to be mutated in a cell line derived from breast cancer that expressed extremely high levels of cyclin E.

Specifications

Antigen FBW7
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene FBXW7
Gene Accession No. Q969H0
Gene Alias AGO, archipelago, Archipelago homolog, archipelago homolog (Drosophila), CDC4, F-box and WD repeat domain containing 7, E3 ubiquitin protein ligase, F-box and WD-40 domain protein 7 (archipelago homolog, Drosophila), F-box and WD-40 domain-containing protein 7, F-box protein FBW7, F-box protein FBX30, F-box protein SEL-10, FBW6, FBW7, FBX30, FBXO30, FBXW6, FLJ11071, hAgo, hCdc4, homolog of C elegans sel-10, SEL10, SEL-10
Gene Symbols FBXW7
Host Species Rabbit
Immunogen Between 50 and 100
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55294
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.