Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FBX09 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$81.70 - $330.00
Specifications
Antigen | FBX09 |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:200-1:1000 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
FBX09 Polyclonal antibody specifically detects FBX09 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
FBX09 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Stem Cell Markers | |
PBS, pH 7.0 | |
26268 | |
IgG | |
Antigen and protein A Affinity-purified |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:200-1:1000 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
Cross-immune reaction antigen 1, dJ341E18.2, F-box only protein 9, F-box protein 9, F-box protein Fbx9, FBX9DKFZp434C0118, KIAA0936, NY-REN-57, Renal carcinoma antigen NY-REN-57, VCIA1 | |
Produced in rabbits immunized with E. coli-derived Human FBX09 fragment | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title