Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FBX10 Rabbit anti-Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | FBX10 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
FBX10 Polyclonal specifically detects FBX10 in Rat samples. It is validated for Western Blot.Specifications
FBX10 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
26267 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Rat | |
F-box protein 10, PRMT11 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human FBX10 (NP_036298.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title